5-Amino-2-methylpyridin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
6-Methylpyridin-3-amin, 3-Amino-6-picolin, 6-Methyl-3-pyridylamin, 5-Amino-2-picolin
M-Clarity™ Qualitätsstufe = MQ100
662704-1G
662704-250MG
SIAL662704-1GEA
36.4
EUR
SIAL662704-1G
SIAL662704-250MG
5-Amino-2-methylpyridin, Sigma-Aldrich®
5-Amino-2-methylpyridin
Formel:
C₆H₈N₂ Molecular Weight: 108,14 g/mol |
MDL:
MFCD00833389 CAS-Nummer: 3430-14-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...