2-Amino-4-methyl-3-nitropyridin 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Methyl-3-nitropyridin-2-amin, 4-Methyl-3-nitro-2-pyridylamin, 2-Amino-3-nitro-4-picolin

L15841.06 L15841.14
L15841.06EA 63.7 EUR
L15841.06 L15841.14
2-Amino-4-methyl-3-nitropyridin 97%
2-Amino-4-methyl-3-nitropyridin

Purity: 97% CAS No.: 6635-86-5 Molecular Formula: C6H7N3O2 Formula Weight: 153.14 Melting Point: 134-138° Beilstein Reference: 139111

Formel: C₆H₇N₃O₂
Molecular Weight: 153,14 g/mol
Schmelzpunkt: 134…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006315
CAS-Nummer: 6635-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR