2-Amino-4-methyl-3-nitropyridin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
4-Methyl-3-nitropyridin-2-amin, 4-Methyl-3-nitro-2-pyridylamin, 2-Amino-3-nitro-4-picolin
M-Clarity™ Qualitätsstufe = MQ100
290084-25G
290084-5G
SIAL290084-25GEA
262
EUR
SIAL290084-25G
SIAL290084-5G
2-Amino-4-methyl-3-nitropyridin, Sigma-Aldrich®
2-Amino-4-methyl-3-nitropyridin
Formel:
C₆H₇N₃O₂ Molecular Weight: 153,14 g/mol Schmelzpunkt: 140 °C |
MDL:
MFCD00006315 CAS-Nummer: 6635-86-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...