4-Ethylpyridin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A12883.18 A12883.30
A12883.18EA 48.4 EUR
A12883.18 A12883.30
4-Ethylpyridin ≥98%
4-Ethylpyridine

Purity: 98% CAS No.: 536-75-4 Molecular Formula: C7H9N Formula Weight: 107.16 Flash Point: 52°(126°F) Refractive Index: 1.5010 Odour: Unpleasant pH: 10 Melting Point: -91° Boiling Point: 167-168° Beilstein Reference: 106481 Merck Reference: 14,3849 UN No.: 1993 MDL No.: MFCD00006450

Formel: C₇H₉N
Molecular Weight: 107.16 g/mol
Siedepunkt: 167…168 °C
Schmelzpunkt: –91 °C
Dichte: 0.942
Flammpunkt: 52 °C (126 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006450
CAS-Nummer: 536-75-4
EINECS: 208-646-7
UN: 1993
ADR: 3 III
Merck Index: 13,03880

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR