3-Benzoylpyridin ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Phenyl-3-pyridylketon

A16695.06 A16695.14 A16695.22
A16695.06EA 35.4 EUR
A16695.06 A16695.14 A16695.22
3-Benzoylpyridin ≥98%
3-Benzoylpyridin

Purity: 98+% CAS No.: 5424-19-1 Molecular Formula: C12H9NO Formula Weight: 183.21 Flash Point: 150°(302°F) Melting Point: 36-40° Boiling Point: 307° Beilstein Reference: 120234 MDL No.: MFCD00006394

Formel: C₁₂H₉NO
Molecular Weight: 183,21 g/mol
Siedepunkt: 307 °C
Schmelzpunkt: 36…40 °C
Flammpunkt: 150 °C (302 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006394
CAS-Nummer: 5424-19-1
EINECS: 226-561-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR