4,4′-Bipyridin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4,4'-Bipyridin, γ,γ'-Dipyridyl
A10187.06
A10187.14
A10187.22
AB110731.0100
A10187.06EA
32.6
EUR
A10187.06
A10187.14
A10187.22
ABCRAB110731.0100
4,4′-Bipyridin ≥98%
4,4'-Bipyridyl
Purity: 98%
CAS No.: 553-26-4
Molecular Formula: C10H8N2
Formula Weight: 156.19
Melting Point: 111-114°
Boiling Point: 304-305°
Merck Reference: 14,3348
UN No.: 2811
MDL No.: MFCD00006416
Formel:
C₁₀H₈N₂ Siedepunkt: 304…305 °C Schmelzpunkt: 111…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006416 CAS-Nummer: 553-26-4 EINECS: 209-036-3 UN: 2811 ADR: 6.1,III Merck Index: 14,3348 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...