4,4′-Bipyridin
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4,4'-Bipyridin, γ,γ'-Dipyridyl
SB01188.1
SB01188.50
MAYBSB01188.1EA
0
EUR
MAYBSB01188.1
MAYBSB01188.50
4,4′-Bipyridin
4,4'-Bipyridyl
Formel:
C₁₀H₈N₂ Molecular Weight: 156.19 g/mol Siedepunkt: 305 Schmelzpunkt: 78 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006416 CAS-Nummer: 553-26-4 EINECS: 209-036-3 UN: 2811 ADR: 6.1 III Merck Index: 13,03383 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...