5-Brompyrimidin 98%

Lieferant: Thermo Scientific
Achtung

204300250 204301000
ACRO204300250EA 58 EUR
ACRO204300250 ACRO204301000
5-Brompyrimidin 98%
5-Bromopyrimidine
Formel: C₄H₃BrN₂
Molecular Weight: 158,99 g/mol
Siedepunkt: 168 °C (727 mmHg)
Schmelzpunkt: 69…73 °C
MDL: MFCD00006117
CAS-Nummer: 4595-59-9
EINECS: 224-992-1

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to beige to light brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 69 to 73 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR