5-Brompyrimidin ≥98%

Lieferant: Thermo Scientific
Achtung

L14172.06 L14172.14 L14172.22
L14172.06EA 24.5 EUR
L14172.06 L14172.14 L14172.22
5-Brompyrimidin ≥98%
5-Bromopyrimidine

Purity: 98% CAS No.: 4595-59-9 Molecular Formula: C4H3BrN2 Formula Weight: 158.99 Melting Point: 69-74° Beilstein Reference: 107326

Formel: C₄H₃BrN₂
Molecular Weight: 158,99 g/mol
Siedepunkt: 168 °C (727 mmHg)
Schmelzpunkt: 69…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006117
CAS-Nummer: 4595-59-9
EINECS: 224-992-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR