2,4-Dichlorpyrimidin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A15131.06 A15131.09 A15131.18 A15131.30
A15131.06EA 44.8 EUR
A15131.06 A15131.09 A15131.18 A15131.30
2,4-Dichlorpyrimidin ≥98%
2,4-Dichloropyrimidine

Purity: 98+% CAS No.: 3934-20-1 Molecular Formula: C4H2Cl2N2 Formula Weight: 148.98 Storage Temperature: 2°C to 8°C Melting Point: 57-62° Boiling Point: 101°/23mm Beilstein Reference: 110911 MDL No.: MFCD00006061

Formel: C₄H₂Cl₂N₂
Siedepunkt: 101 °C (23 mmHg)
Schmelzpunkt: 57…62 °C
MDL: MFCD00006061
CAS-Nummer: 3934-20-1
EINECS: 223-508-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR