2-Chlorpyrimidin ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A15394.09
A15394.14
A15394.22
A15394.09EA
35.8
EUR
A15394.09
A15394.14
A15394.22
2-Chlorpyrimidin ≥98%
2-Chloropyrimidine
Purity: 98%
CAS No.: 1722-12-9
Molecular Formula: C4H3ClN2
Formula Weight: 114.54
Flash Point: 98°(208°F)
Melting Point: 64-67°
Boiling Point: 75-76°/10mm
Beilstein Reference: 107171
UN No.: 2811
MDL No.: MFCD00006060
Formel:
C₄H₃ClN₂ Molecular Weight: 114.53 g/mol Siedepunkt: 75…76 °C (10 mmHg) Schmelzpunkt: 64…67 °C Flammpunkt: 98 °C (208 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006060 CAS-Nummer: 1722-12-9 EINECS: 217-020-2 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...