2,4-Dichlor-5-fluorpyrimidin
Lieferant: CHEMPUR
Synonyme:
5-Fluoro-2,4-dichloropyrimidine
FL10972.1
010972.5G
010972-25G
010972-100G
CHMPFL10972.1EA
0
EUR
CHMPFL10972.1
FLUO010972.5G
FLUO010972-25G
FLUO010972-100G
2,4-Dichlor-5-fluorpyrimidin
2,4-Dichlor-5-fluorpyrimidin
Formel:
C₄HCl₂FN₂ Molecular Weight: 166.97 g/mol |
MDL:
MFCD00233551 CAS-Nummer: 2927-71-1 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...