2,4,6-Trifluorpyrimidin ≥98%

Lieferant: Thermo Scientific
Gefahr

L04404.03 L04404.06
L04404.03EA 53 EUR
L04404.03 L04404.06
2,4,6-Trifluorpyrimidin ≥98%
2,4,6-Trifluoropyrimidine

Purity: 98% CAS No.: 696-82-2 Molecular Formula: C4HF3N2 Formula Weight: 134.06 Refractive Index: 1.4050 Boiling Point: 100-102° UN No.: 3265 MDL No.: MFCD00039705

Formel: C₄HF₃N₂
Molecular Weight: 134,06 g/mol
Siedepunkt: 100…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039705
CAS-Nummer: 696-82-2
EINECS: 211-801-1
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR