4-Aminopyrimidin

Lieferant: CHEMPUR
Achtung

FL13417.POR 013417-1G 013417-5G
CHMPFL13417.POREA 0 EUR
CHMPFL13417.POR FLUO013417-1G FLUO013417-5G
4-Aminopyrimidin
4-Aminopyrimidin
Formel: C₄H₅N₃
Molecular Weight: 95.1 g/mol
Schmelzpunkt: 154…156 °C
MDL: MFCD00006112
CAS-Nummer: 591-54-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR