2,4-Diaminopyrimidin, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Pyrimidin-2,4-diyltriamin

M-Clarity™ Qualitätsstufe = MQ100

468231-1G 468231-5G
SIAL468231-1GEA 76.4 EUR
SIAL468231-1G SIAL468231-5G
2,4-Diaminopyrimidin, Sigma-Aldrich®
2,4-Diaminopyrimidin
Formel: C₄H₆N₄
Molecular Weight: 110,12 g/mol
MDL: MFCD00038023
CAS-Nummer: 156-81-0
EINECS: 205-862-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR