3-Acetylpyrrol ≥97%
Lieferant: Thermo Scientific
A12073.03
A12073.06
A12073.03EA
82.1
EUR
A12073.03
A12073.06
3-Acetylpyrrol ≥97%
3-Acetylpyrrol
Purity: 97%
CAS No.: 1072-82-8
Molecular Formula: C6H7NO
Formula Weight: 109.13
Odour: Odourless
Melting Point: 110-114°
Beilstein Reference: 107151
MDL No.: MFCD00067759
Formel:
C₆H₇NO Molecular Weight: 109,13 g/mol Schmelzpunkt: 110…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067759 CAS-Nummer: 1072-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...