1-Methylpyrrol ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
N-Methylpyrrol
B21451.AE
B21451.AP
B21451.AEEA
70.3
EUR
B21451.AE
B21451.AP
1-Methylpyrrol ≥99%
1-Methylpyrrol
Purity: 99%
CAS No.: 96-54-8
Molecular Formula: C5H7N
Formula Weight: 81.12
Flash Point: 15°(59°F)
Refractive Index: 1.4880
Melting Point: -57°
Boiling Point: 112-113°
Beilstein Reference: 104181
UN No.: 1993
MDL No.: MFCD00005345
Formel:
C₅H₇N Molecular Weight: 81,12 g/mol Siedepunkt: 112…113 °C Schmelzpunkt: –57 °C Dichte: 0,912 Flammpunkt: 15 °C (59 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005345 CAS-Nummer: 96-54-8 EINECS: 202-513-7 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...