1-Phenylpyrrol ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: N-Phenylpyrrol

L07973.06 L07973.14
L07973.06EA 43.1 EUR
L07973.06 L07973.14
1-Phenylpyrrol ≥99%
1-Phenylpyrrol

Purity: 99% CAS No.: 635-90-5 Molecular Formula: C10H9N Formula Weight: 143.19 Melting Point: 57-60° Boiling Point: 131°/30mm MDL No.: MFCD00005343

Formel: C₁₀H₉N
Molecular Weight: 143.19 g/mol
Siedepunkt: 131 °C (30 mmHg)
Schmelzpunkt: 57…60 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005343
CAS-Nummer: 635-90-5
EINECS: 211-242-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR