1,1,3,3-Tetramethyldisilazan ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Bis(dimethylsilyl)amin, TMDS, N-(Dimethylsilyl)-1,1-dimethylsilylamin
A14304.09
A14304.18
A14304.09EA
68.3
EUR
A14304.09
A14304.18
1,1,3,3-Tetramethyldisilazan ≥97%
1,1,3,3-Tetramethyldisilazan
Purity: 97%
CAS No.: 15933-59-2
Molecular Formula: C4H15NSi2
Formula Weight: 133.34
Flash Point: -3°(26°F)
Refractive Index: 1.4055
Odour: Ammonia-like
Boiling Point: 99-100°
Beilstein Reference: 741869
UN No.: 2924
MDL No.: MFCD00025626
Formel:
C₄H₁₅NSi₂ Molecular Weight: 133.34 g/mol Siedepunkt: 99…100 °C Dichte: 0.768 Flammpunkt: –3 °C (26 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00025626 CAS-Nummer: 15933-59-2 EINECS: 240-072-2 UN: 2924 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...