1,3-Dibutyl-1,1,3,3-tetramethyldisilazan

Lieferant: CHEMPUR SILANE

SILS05925.5 SILS05925.25 SILS05925.100
SILNSILS05925.5EA 0 EUR
SILNSILS05925.5 SILNSILS05925.25 SILNSILS05925.100
1,3-Dibutyl-1,1,3,3-tetramethyldisilazan
1,3-Dibutyl-1,1,3,3-tetramethyldisilazan
Formel: C₁₂H₃₁NSi₂
Molecular Weight: 245.56 g/mol
Siedepunkt: 96
Dichte: 0.8 g/cm³ (20 °C)
CAS-Nummer: 82356-80-7
UN: 3267
ADR: 8 I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR