1,3-Dioctadecyl-1,1,3,3-tetramethyldisilazan

Lieferant: CHEMPUR SILANE

SILS07662.10 SILS07662.25 SILS07662.100
SILNSILS07662.10EA 0 EUR
SILNSILS07662.10 SILNSILS07662.25 SILNSILS07662.100
1,3-Dioctadecyl-1,1,3,3-tetramethyldisilazan
1,3-Dioctadecyl-1,1,3,3-tetramethyldisilazan
Formel: C₄₀H₈₇NSi₂
CAS-Nummer: 82356-82-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR