1,3-Dioctyl-1,1,3,3-tetramethyldisilazan

Lieferant: CHEMPUR SILANE

SILS07700.10 SILS07700.25 SILS07700.100
SILNSILS07700.10EA 0 EUR
SILNSILS07700.10 SILNSILS07700.25 SILNSILS07700.100
1,3-Dioctyl-1,1,3,3-tetramethyldisilazan
1,3-Dioctyl-1,1,3,3-tetramethyldisilazan
Formel: C₂₀H₄₇NSi₂
CAS-Nummer: 69519-51-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR