Heptamethyldisilazan
Lieferant: CHEMPUR SILANE
|
Gefahr
|
SILS09250.10
SILS09250.25
SILS09250.100
SILNSILS09250.10EA
0
EUR
SILNSILS09250.10
SILNSILS09250.25
SILNSILS09250.100
Heptamethyldisilazan
Heptamethyldisilazan
Formel:
C₇H₂₁NSi₂ Siedepunkt: 148 °C (1013 hPa) Dichte: 0,79 g/cm³ (20 °C) |
MDL:
MFCD00008262 CAS-Nummer: 920-68-3 EINECS: 213-061-5 UN: 2921 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...