Pentamethyldisiloxan

Lieferant: CHEMPUR SILANE

Synonyme: 1,1,1,3,3-Pentamethyldisiloxane

SILS12900.5 SILS12900.25 SILS12900.100
SILNSILS12900.5EA 0 EUR
SILNSILS12900.5 SILNSILS12900.25 SILNSILS12900.100
Pentamethyldisiloxan
Pentamethyldisiloxane
Formel: C₅H₁₆OSi₂
Molecular Weight: 148.35 g/mol
Siedepunkt: 82
Dichte: 0.7588 g/cm³ (20 °C)
MDL: MFCD00053658
CAS-Nummer: 1438-82-0
EINECS: 215-873-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR