1,1,3,3,5,5-Hexamethyltrisiloxan
Lieferant: CHEMPUR SILANE
Synonyme:
1H,5H-Hexamethyltrisiloxane
SILS09700.10
SILS09700.25
SILS09700.100
SILNSILS09700.10EA
0
EUR
SILNSILS09700.10
SILNSILS09700.25
SILNSILS09700.100
1,1,3,3,5,5-Hexamethyltrisiloxan
1,1,3,3,5,5-Hexamethyltrisiloxan
Formel:
C₆H₂₀O₂Si₃ Molecular Weight: 208.48 g/mol Siedepunkt: 128 Schmelzpunkt: –67 °C Dichte: 0.8246 g/cm³ (20 °C) |
MDL:
MFCD00039788 CAS-Nummer: 1189-93-1 EINECS: 214-716-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...