1,1,1,3,3,5,5-Heptamethyltrisiloxan

Lieferant: CHEMPUR SILANE

SILS09275.10 SILS09275.25 SILS09275.100
SILNSILS09275.10EA 0 EUR
SILNSILS09275.10 SILNSILS09275.25 SILNSILS09275.100
1,1,1,3,3,5,5-Heptamethyltrisiloxan
1,1,1,3,3,5,5-Heptamethyltrisiloxan
Formel: C₇H₂₂O₂Si₃
Molecular Weight: 222,51 g/mol
MDL: MFCD00054788
CAS-Nummer: 2895-07-0
EINECS: 220-774-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR