1,1,1,3,5,5,5-Heptamethyltrisiloxan

Lieferant: CHEMPUR SILANE

Synonyme: 3H-Heptamethyltrisiloxan

SILS09300.5 SILS09300.25 SILS09300.100
SILNSILS09300.5EA 0 EUR
SILNSILS09300.5 SILNSILS09300.25 SILNSILS09300.100
1,1,1,3,5,5,5-Heptamethyltrisiloxan
1,1,1,3,5,5,5-Heptamethyltrisiloxan
Formel: C₇H₂₂O₂Si₃
Molecular Weight: 222,51 g/mol
MDL: MFCD00048000
CAS-Nummer: 1873-88-7
EINECS: 217-496-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR