1,1,1,3,5,7,7,7-Octamethyltetrasiloxan 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3H,5H-Octamethyltetrasiloxane

L16956.06 L16956.14
L16956.06EA 45.9 EUR
L16956.06 L16956.14
1,1,1,3,5,7,7,7-Octamethyltetrasiloxan 96%
1,1,1,3,5,7,7,7-Octamethyltetrasiloxan

Purity: 96% CAS No.: 16066-09-4 Molecular Formula: C8H26O3Si4 Formula Weight: 282.64 Flash Point: 78°(172°F) Refractive Index: 1.3860 Boiling Point: 169° MDL No.: MFCD00053664

Formel: C₈H₂₆O₃Si₄
Molecular Weight: 282.63 g/mol
Siedepunkt: 169 °C
Dichte: 0.863
Flammpunkt: 78 °C (172 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00053664
CAS-Nummer: 16066-09-4
EINECS: 240-209-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR