1,1,1,3,5,7,7,7-Octamethyltetrasiloxan

Lieferant: CHEMPUR SILANE

Synonyme: 3H,5H-Octamethyltetrasiloxane

SILS12425.5 SILS12425.25
SILNSILS12425.5EA 0 EUR
SILNSILS12425.5 SILNSILS12425.25
1,1,1,3,5,7,7,7-Octamethyltetrasiloxan
1,1,1,3,5,7,7,7-Octamethyltetrasiloxan
Formel: C₈H₂₆O₃Si₄
Molecular Weight: 282.63 g/mol
MDL: MFCD00053664
CAS-Nummer: 16066-09-4
EINECS: 240-209-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR