1,1,3,3,5,5,7,7-Octamethyltetrasiloxan

Lieferant: CHEMPUR SILANE

SILS12450.10 SILS12450.25 SILS12450.100
SILNSILS12450.10EA 0 EUR
SILNSILS12450.10 SILNSILS12450.25 SILNSILS12450.100
1,1,3,3,5,5,7,7-Octamethyltetrasiloxan
1,1,3,3,5,5,7,7-Octamethyltetrasiloxan
Formel: C₈H₂₆O₃Si₄
Molecular Weight: 282.63 g/mol
MDL: MFCD00039789
CAS-Nummer: 1000-05-1
EINECS: 213-669-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR