3-Brom-2,5-dichlorthiophen ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

A10603.03 A10603.06 A10603.14
A10603.03EA 20.7 EUR
A10603.03 A10603.06 A10603.14
3-Brom-2,5-dichlorthiophen ≥97%
3-Bromo-2,5-dichlorothiophene

Purity: 97% CAS No.: 60404-18-4 Molecular Formula: C4HBrCl2S Formula Weight: 231.93 Refractive Index: 1.6080 Boiling Point: 95-97°/14mm MDL No.: MFCD00041440

Formel: C₄HBrCl₂S
Molecular Weight: 231.93 g/mol
Siedepunkt: 95…97 °C (14 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00041440
CAS-Nummer: 60404-18-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR