3-Brom-2,5-dichlorthiophen

Lieferant: Thermo Scientific

CD09413.1 CD09413.10 CD09413.50
MAYBCD09413.1EA 0 EUR
MAYBCD09413.1 MAYBCD09413.10 MAYBCD09413.50
3-Brom-2,5-dichlorthiophen
3-Bromo-2,5-dichlorothiophene
Formel: C₄HBrCl₂S
Molecular Weight: 231.93 g/mol
Siedepunkt: 89…92 °C (11 mmHg)
MDL: MFCD00041440
CAS-Nummer: 60404-18-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR