4-Brom-2-methylthiophen

Lieferant: FLUOROCHEM

022016-5G 022016-25G
FLUO022016-5GEA 55.4 EUR
FLUO022016-5G FLUO022016-25G
4-Brom-2-methylthiophen
4-Bromo-2-methylthiophene
Formel: C₅H₅BrS
Molecular Weight: 177,06 g/mol
MDL: MFCD04116367
CAS-Nummer: 29421-92-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR