4-Brom-2-methylthiophen, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

648728-1G 648728-5G
SIAL648728-1GEA 37.7 EUR
SIAL648728-1G SIAL648728-5G
4-Brom-2-methylthiophen, Sigma-Aldrich®
4-Bromo-2-methylthiophene
Formel: C₅H₅BrS
Molecular Weight: 177,06 g/mol
MDL: MFCD04116367
CAS-Nummer: 29421-92-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR