2-Chlor-5-methylthiophen ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
B21657.06
B21657.14
B21657.06EA
65.4
EUR
B21657.06
B21657.14
2-Chlor-5-methylthiophen ≥97%
2-Chloro-5-methylthiophene
Purity: 97%
CAS No.: 17249-82-0
Molecular Formula: C5H5ClS
Formula Weight: 132.61
Flash Point: 47°(117°F)
Refractive Index: 1.5356
Odour: Characteristic
Boiling Point: 155-156°
UN No.: 1992
MDL No.: MFCD01632145
Formel:
C₅H₅ClS Siedepunkt: 155…156 °C Dichte: 1,21 Flammpunkt: 47 °C (117 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01632145 CAS-Nummer: 17249-82-0 UN: 1992 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...