2-Chlor-6-methylthiophen 96%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Chloro-6-methylthiophenol
L10499.06
L10499.03
L10499.14
L10499.06EA
162
EUR
L10499.06
L10499.03
L10499.14
2-Chlor-6-methylthiophen 96%
2-Chloro-6-methylthiophene
Purity: 96%
CAS No.: 18858-05-4
Molecular Formula: C7H7ClS
Formula Weight: 158.65
Refractive Index: 1.5950
Boiling Point: 135-136°/35mm
MDL No.: MFCD00041422
Formel:
C₅H₅ClS Molecular Weight: 132,61 g/mol Siedepunkt: 135…136 °C (35 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041422 CAS-Nummer: 18858-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...