2-Benzoylthiophen ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: Phenyl-2-thienylketon

A12036.06 A12036.09 A12036.18
A12036.06EA 25.6 EUR
A12036.06 A12036.09 A12036.18
2-Benzoylthiophen ≥98%
2-Benzoylthiophen

Purity: 98% CAS No.: 135-00-2 Molecular Formula: C11H8OS Formula Weight: 188.25 Melting Point: 55-57° Boiling Point: 124-126°/0.5mm MDL No.: MFCD00022495

Formel: C₁₁H₈OS
Molecular Weight: 188.25 g/mol
Siedepunkt: 124…126 °C (0.5 mmHg)
Schmelzpunkt: 55…57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00022495
CAS-Nummer: 135-00-2
EINECS: 205-169-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR