1-Brom-3,5-dimethyladamantan, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Brom-3,5-dimethyltricyclo[3.3.1.1(3,7)]decan
M-Clarity™ Qualitätsstufe = MQ100
187844-25G
187844-5G
SIAL187844-25GEA
68.2
EUR
SIAL187844-25G
SIAL187844-5G
1-Brom-3,5-dimethyladamantan, Sigma-Aldrich®
1-Brom-3,5-dimethyladamantan
Formel:
C₁₂H₁₉Br Molecular Weight: 243,19 g/mol |
MDL:
MFCD00077197 CAS-Nummer: 941-37-7 EINECS: 213-378-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...