Streptomycinsulfat USP, Molekularbiologische Qualität

Lieferant: G Bioscienes
Gefahr

GENORC-197EA 208 EUR
GENORC-197 GENORC-196
Streptomycinsulfat USP, Molekularbiologische Qualität
Streptomycin sulphate

Mode of action and anti-microbial spectrum streptomycin sulphate is an aminoglycoside antibiotic and has a bactericidal action against many gram-negative bacteria. It binds to the #12 protein on the ribosomal 30S subunit and inhibits protein synthesis.

Formel: C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
Molecular Weight: 1457,4 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00037023
CAS-Nummer: 3810-74-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR