Lithiumchlorid ≥99%, weißes kristallines Pulver für die Molekularbiologie
Lieferant: MP Biomedicals
Achtung
0219401080 0219401090
ICNA0219401080EA 0 EUR
ICNA0219401080 ICNA0219401090
Lithiumchlorid ≥99%, weißes kristallines Pulver für die Molekularbiologie
Lithium chloride
Lithium Chloride is a metal halide utilised in a variety of assays to study cell-fate, neurobiology and antiviral properties.
Lithium Chloride is used as an electrolyte for low temperature dry battery cells and as an oxidation catalyst. It is a solubilizer for polyamides and cellulose when used with amide solvents, and is a chlorinating agent for steroid substrates. It has also been utilized in: Large scale plasmid DNA isolation without ultracentrifugation; Protein extraction and protein crystallization; Crystallization of other biological structures, including vitamin B12-RNA aptamer and the L-A virus particle; Inhibits the expression and secretion of insulin-like growth factor-binding protein-1 in H4-II-E cells; Used in the synthesis of beta-substituted alpha-amino acid derivatives; May be used to selectively precipitate RNA.
Formel: LiCl
Molecular Weight: 42.39 g/mol
Siedepunkt: 1382
Schmelzpunkt: 614
Lagertemperatur: Raumtemperatur
MDL: MFCD00011078
CAS-Nummer: 7447-41-8
Merck Index: 13,05550
Bestellen
Spezifikation Testergebnisse
Identity Test Passes
Purity >99%