Potassium-L-aspartate ≥98% (durch HPLC), weißes Pulver, Sigma-Aldrich®

Lieferant: Merck

Synonyme: L-Aspartic Acid Potassium Salt, L-Aspartic Acid Monopotassium Salt, L-Asparaginic Acid Potassium Salt

M-Clarity™ Qualitätsstufe = MQ200

A6558-100G A6558-1KG A6558-25G
SIALA6558-100GEA 199 EUR
SIALA6558-100G SIALA6558-1KG SIALA6558-25G
Potassium-L-aspartate ≥98% (durch HPLC), weißes Pulver, Sigma-Aldrich®
Potassium-L-aspartate
Formel: C₄H₆NO₄K
Molecular Weight: 171,19 g/mol
MDL: MFCD00021083
CAS-Nummer: 1115-63-5

Bestellen

Spezifikation Testergebnisse

Water (by Karl Fischer) ≤6%
Nitrogen (anhydrous) 8.0 - 8.4%
Potassium (K) anhydrous 21.0 - 24.6%
Purity (HPLC) ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR