Aminophylline, wasserfrei 98%, Pulver

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3,7-Dihydro-1,3-dimethylpurine-2,6-dione, complex with 1,2-ethanediamine (2:1), 1,3-Dimethyl-3,7-dihydro-1H-purine-2,6-dione hemiethane-1,2-diamine salt

J60705.14 J60705.22
J60705.14EA 44.6 EUR
J60705.14 J60705.22
Aminophylline, wasserfrei 98%, Pulver
Aminophylline

A non-selective phosphodiesterase inhibitor


Aminophylline exhibits vasodilatory and anti-inflammatory activity and is occasionally clinically used to treat chronic obstructive pulmonary disorder (COPD).


In animal models of allergen-induced lung inflammation, aminophylline decreases levels of IL-5, IL-8, and eosinophils. It is mostly used to prepare solutions for intravenous injections.

Formel: C₇H₈N₄O₂
Molecular Weight: 420,43 g/mol
Schmelzpunkt: 269…274 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00013221
CAS-Nummer: 317-34-0
EINECS: 206-264-5
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Purity 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR