Poly(ethylene glycol) methacrylate inhibiert, Sigma-Aldrich®
Lieferant: Merck
Achtung
M-Clarity™ Qualitätsstufe = MQ100
409529-100ML 409529-500ML
SIAL409529-100MLEA 95.5 EUR
SIAL409529-100ML SIAL409529-500ML
Poly(ethylene glycol) methacrylate inhibiert, Sigma-Aldrich®
Poly(ethylene glycol) methacrylate
Comonomer in emulsion polymerization, thickener and dispersant and suspending aid.
  • Copolymerizable, non-ionic stabilizer
  • Hydrophilic, modifying comonomer and cross-linking agent
Average Mn 500, contains 900 ppm monomethyl ether hydroquinone as inhibitor
Formel: H₂C=C(CH₃)CO(OCH₂CH₂)nOH
Dichte: 1,101 g/mL (25 °C)
Flammpunkt: 113 °C
MDL: MFCD00081879
CAS-Nummer: 25736-86-1