Anti-CALB1 Mouse Monoclonal Antibody [clone: 4H7]

Lieferant: Biosensis
BSENM-1797-100EA 302 EUR
BSENM-1797-100
Anti-CALB1 Mouse Monoclonal Antibody [clone: 4H7]
Antikörper
Buffers cytosolic calcium. May stimulate a membrane Ca2+-ATPase and a 3',5'-cyclic nucleotide phosphodiesterase. Ref: uniprot.org

Western blotting (1:1,000-1:5,000) and Immunohistochemistry (1:1,000). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CALB1
Clonality: Monoclonal
Clone: 4H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Cow, Pig, Horse, Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR