Aminophylline ≥98%, kristalliner Feststoff
Lieferant: Cayman Chemical
|
Gefahr
|
Synonyme:
3,7-Dihydro-1,3-dimethylpurine-2,6-dione, complex with 1,2-ethanediamine (2:1), 1,3-Dimethyl-3,7-dihydro-1H-purine-2,6-dione hemiethane-1,2-diamine salt
Aminophylline is a competitive and non-selective phosphodiesterase inhibitor (IC₅₀=120 µM) and adenosine receptor antagonist. It is a complex of theophylline and ethylenediamine that has in vivo bronchodilator and vasodilator effects.
Aminophylline suppresses maternal separation- and acetic acid administration-induced visceral hypersensitivity to colorectal distension in a rat model of irritable bowel syndrome with diarrhea (IBS-D). Aminophylline also provides renoprotection against murine renal ischemia-reperfusion injury.
Bitte beachten Not for human or veterinary use.
Formel:
C₇H₈N₄O₂ Molecular Weight: 420.43 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013221 CAS-Nummer: 317-34-0 UN: 2811 ADR: 6.1 III |
Spezifikation Testergebnisse
Purity | ≥98% |
λmax | 272 nm |
Stability | ≥2 years |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...