Cyclopamine (aus V. Californicum) ≥97% (durch HPLC), Millipore®
Lieferant: Merck Millipore Calbiochem
Achtung
Synonyme: (2'R,3S,3'R,3a'S,6aS,6bS,6'S,7a'R,11aS,11bR)-3',6',10,11b-tetramethyl-1,2,3,3a',4,4',5',6,6a,6b,6',7,7',7a',8,11,11a,11b-octadecahydro-3'H-spiro[benzo[a]fluorene-9,2'-furo[3,2-b]pyridin]-3-ol
239803-1 239803-5MG
239803-1EA 280 EUR
239803-1 239803-5MG
Cyclopamine (aus V. Californicum) ≥97% (durch HPLC), Millipore®
Cyclopamine
Cyclopamine, V. californicum, is a cell-permeable steroidal alkaloid & cholesterol mimic that specifically antagonises sonic hedgehog signaling through direct interaction with Smo.
Verpackung: Packaged under inert gas.
Formel: C₂₇H₄₁NO₂
Molecular Weight: 411,63 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD09878269
CAS-Nummer: 4449-51-8