Potassium bis(oxalato)oxotitanate(IV) dihydrate ≥98% (Ti-Basis), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Dipotassium oxodioxalatotitanate(IV) dihydrate, Potassium titanium oxide oxalate dihydrate
M-Clarity™ Qualitätsstufe = MQ100
14007-1KG
14007-6X1KG
SIAL14007-1KGEA
102
EUR
SIAL14007-1KG
SIAL14007-6X1KG
Potassium bis(oxalato)oxotitanate(IV) dihydrate ≥98% (Ti-Basis), Sigma-Aldrich®
Potassium bis(oxalato)oxotitanate(IV) dihydrate
Formel:
K₂[TiO(C₂O₄)₂]·2H₂O Molecular Weight: 354.10 g/mol (318.10 anhy) |
MDL:
MFCD00150592 CAS-Nummer: 14402-67-6 |
Spezifikation Testergebnisse
Assay (Ti basis) | ≥98% |
Iron (Fe) | ≥98% |
Potassium (K) | ≤26% |
Titration with KMnO₄ | ≥90.0% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...