Ticagrelor ≥98%

Lieferant: Cayman Chemical

Synonyme: AZD 6140, AR-C 126532XX

15425-10 15425-5 15425-25
CAYM15425-10EA 67 EUR
CAYM15425-10 CAYM15425-5 CAYM15425-25
Ticagrelor ≥98%
Ticagrelor

Ticagrelor is a reversible antagonist of the platelet purinergic P2Y12 receptor (Ki = 14 nM; IC50 = 1.8 μM), which is the main receptor responsible for ADP-induced platelet aggregation. It functions by directly changing the conformation of the P2Y12 receptor to inhibit ADP binding. Formulations containing ticagrelor have been used to reduce the rate of thrombotic cardiovascular events in patients with acute coronary syndrome.

Formel: C₂₃H₂₈F₂N₆O₄S
Molecular Weight: 522,6 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD09954148
CAS-Nummer: 274693-27-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR