Ethyl-1-(hydroxymethyl)cyclopropanecarboxylate 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
Ethyl 1-(hydroxymethyl)cyclopropanecarboxylate
H62277.03
H62277.06
H62277.03EA
232
EUR
H62277.03
H62277.06
Ethyl-1-(hydroxymethyl)cyclopropanecarboxylate 95%
Ethyl-1-(hydroxymethyl)cyclopropanecarboxylate
Ethyl 1-(hydroxymethyl)cyclopropanecarboxylate acts as a ligand for Suzuki coupling reaction.
Formel:
C₇H₁₂O₃ Molecular Weight: 144.17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09759206 CAS-Nummer: 3697-68-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...