Diethylpyrocarbonat ≥97%, ultrarein

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Diethyldicarbonat, Pyrokohlensäurediethylester, DEPC

J14710.AE J14710.AC
J14710.AEEA 415 EUR
J14710.AE J14710.AC
Diethylpyrocarbonat ≥97%, ultrarein
Diethylpyrocarbonat

A strong, non-specific inhibitor of ribonuclease. Reacts with the N1 of histidine residues in proteins under mild conditions. It can be used to cross-link and modify proteins by ethoxyformylation of NH₂ and imidazole groups in proteins. It reacts with amines and thus cannot be used with Tris buffer.

Formel: O(COOC₂H₅)₂
Molecular Weight: 162.14 g/mol
Siedepunkt: 93…94 °C (18 mmHg)
Schmelzpunkt: 69 °C
Dichte: 1.101
Flammpunkt: 69 °C (156 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00009106
CAS-Nummer: 1609-47-8
EINECS: 216-542-8
Merck Index: 14,07998

Bestellen

Spezifikation Testergebnisse

Assay (unspecified) ≥97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR